3-[(4-Methoxybenzyl)oxy]pyrrolidine

Supplier: Matrix Scientific

050872-500MG 050872-2.5G
101893-384EA 364.62 USD
101893-384 101893-386
3-[(4-Methoxybenzyl)oxy]pyrrolidine
3-[(4-Methoxybenzyl)oxy]pyrrolidine

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD16709433
CAS Number: 933759-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR