4-{[(4-Methoxybenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050871-500MG 050871-2.5G
101893-380EA 438.46 USD
101893-380 101893-382
4-{[(4-Methoxybenzyl)oxy]methyl}piperidine hydrochloride
4-{[(4-Methoxybenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₂
MDL Number: MFCD13561251
CAS Number: 1219949-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR