3-{[(4-Methoxybenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050870-500MG 050870-2.5G
101893-376EA 364.62 USD
101893-376 101893-378
3-{[(4-Methoxybenzyl)oxy]methyl}piperidine hydrochloride
3-{[(4-Methoxybenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₂
MDL Number: MFCD13561250
CAS Number: 1219977-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR