4-[(4-Methoxybenzyl)oxy]piperidine

Supplier: Matrix Scientific

Synonyms: 4-[(4-methoxyphenyl)methoxy]piperidine

050869-500MG 050869-2.5G
101893-372EA 753.85 USD
101893-372 101893-374
4-[(4-Methoxybenzyl)oxy]piperidine
4-[(4-Methoxybenzyl)oxy]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MW: 221.3 g/mol
MDL Number: MFCD11644280
CAS Number: 1220028-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR