3-[(3,5-Dimethoxybenzyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050867-500MG 050867-2.5G
101893-364EA 364.62 USD
101893-364 101893-366
3-[(3,5-Dimethoxybenzyl)oxy]pyrrolidine hydrochloride
3-[(3,5-Dimethoxybenzyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₃
MDL Number: MFCD13561247
CAS Number: 1220036-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR