3,5-Dimethoxybenzyl-4-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050866-500MG 050866-2.5G
101893-360EA 364.62 USD
101893-360 101893-362
3,5-Dimethoxybenzyl-4-piperidinylmethyl ether hydrochloride
3,5-Dimethoxybenzyl-4-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₃
MDL Number: MFCD13561246
CAS Number: 1220020-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR