3,5-Dimethoxybenzyl-3-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050865-500MG 050865-2.5G
101893-356EA 364.62 USD
101893-356 101893-358
3,5-Dimethoxybenzyl-3-piperidinylmethyl ether hydrochloride
3,5-Dimethoxybenzyl-3-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₃
MDL Number: MFCD13561245
CAS Number: 1219977-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR