3-[(3,5-Dimethoxybenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050863-500MG 050863-2.5G
101893-348EA 364.62 USD
101893-348 101893-350
3-[(3,5-Dimethoxybenzyl)oxy]piperidine hydrochloride
3-[(3,5-Dimethoxybenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₃
MDL Number: MFCD13561243
CAS Number: 1220034-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR