3-[(3,5-Difluorobenzyl)oxy]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050862-500MG
050862-2.5G
101893-344EA
364.62
USD
101893-344
101893-346
3-[(3,5-Difluorobenzyl)oxy]pyrrolidine hydrochloride
3-[(3,5-Difluorobenzyl)oxy]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄ClF₂NO |
MDL Number:
MFCD13561242 CAS Number: 1220028-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...