3-{[(3,5-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050860-500MG 050860-2.5G
101893-336EA 364.62 USD
101893-336 101893-338
3-{[(3,5-Difluorobenzyl)oxy]methyl}piperidine hydrochloride
3-{[(3,5-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₂NO
MDL Number: MFCD13561240
CAS Number: 1220020-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR