4-[(3,5-Difluorobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050859-500MG 050859-2.5G
101893-332EA 364.62 USD
101893-332 101893-334
4-[(3,5-Difluorobenzyl)oxy]piperidine hydrochloride
4-[(3,5-Difluorobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClF₂NO
MDL Number: MFCD13561239
CAS Number: 1220016-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR