3-[(3,5-Difluorobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050858-500MG 050858-2.5G
101893-328EA 364.62 USD
101893-328 101893-330
3-[(3,5-Difluorobenzyl)oxy]piperidine hydrochloride
3-[(3,5-Difluorobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClF₂NO
MDL Number: MFCD13561238
CAS Number: 1220020-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR