3-[(3-Methoxybenzyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050857-500MG 050857-2.5G
101893-324EA 364.62 USD
101893-324 101893-326
3-[(3-Methoxybenzyl)oxy]pyrrolidine hydrochloride
3-[(3-Methoxybenzyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO₂
MDL Number: MFCD13561237
CAS Number: 1220034-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR