4-{[(3-Methoxybenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050856-500MG 050856-2.5G
101893-320EA 364.62 USD
101893-320 101893-322
4-{[(3-Methoxybenzyl)oxy]methyl}piperidine hydrochloride
4-{[(3-Methoxybenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₂
MDL Number: MFCD13561236
CAS Number: 1219982-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR