4-{[(2,6-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050851-500MG 050851-2.5G
101893-300EA 364.62 USD
101893-300 101893-302
4-{[(2,6-Difluorobenzyl)oxy]methyl}piperidine hydrochloride
4-{[(2,6-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₂NO
MDL Number: MFCD13561231
CAS Number: 1220020-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR