3-{[(2,6-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050850-500MG 050850-2.5G
101893-296EA 364.62 USD
101893-296 101893-298
3-{[(2,6-Difluorobenzyl)oxy]methyl}piperidine hydrochloride
3-{[(2,6-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₂NO
MDL Number: MFCD13561230
CAS Number: 1220020-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR