4-[(2,6-Difluorobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050849-500MG 050849-2.5G
101893-292EA 530.77 USD
101893-292 101893-294
4-[(2,6-Difluorobenzyl)oxy]piperidine hydrochloride
4-[(2,6-Difluorobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClF₂NO
MDL Number: MFCD13561229
CAS Number: 1220035-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR