4-{[(2,4-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050846-500MG 050846-2.5G
101893-280EA 364.62 USD
101893-280 101893-282
4-{[(2,4-Difluorobenzyl)oxy]methyl}piperidine hydrochloride
4-{[(2,4-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₂NO
MDL Number: MFCD13561226
CAS Number: 1220036-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR