3-{[(2,4-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050845-500MG 050845-2.5G
101893-276EA 364.62 USD
101893-276 101893-278
3-{[(2,4-Difluorobenzyl)oxy]methyl}piperidine hydrochloride
3-{[(2,4-Difluorobenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₂NO
MDL Number: MFCD13561225
CAS Number: 1220034-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR