3-[(2,4-Difluorobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050843-500MG 050843-2.5G
101893-268EA 364.62 USD
101893-268 101893-270
3-[(2,4-Difluorobenzyl)oxy]piperidine hydrochloride
3-[(2,4-Difluorobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClF₂NO
MDL Number: MFCD13561223
CAS Number: 1220020-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR