4-Iodobenzyl-4-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050841-500MG 050841-2.5G
101893-260EA 364.62 USD
101893-260 101893-262
4-Iodobenzyl-4-piperidinylmethyl ether hydrochloride
4-Iodobenzyl-4-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClINO
MDL Number: MFCD13561221
CAS Number: 1220020-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR