4-[(4-Iodobenzyl)oxy]piperidine hydrochloride
Supplier: Matrix Scientific
Synonyms:
4-((4-Iodobenzyl)oxy)piperidine hydrochloride
050839-500MG
050839-2.5G
101893-252EA
364.62
USD
101893-252
101893-254
4-[(4-Iodobenzyl)oxy]piperidine hydrochloride
4-[(4-Iodobenzyl)oxy]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClINO MW: 353.63 g/mol |
MDL Number:
MFCD13561219 CAS Number: 1220034-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...