3-[(4-Bromobenzyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050837-500MG 050837-2.5G
101893-244EA 364.62 USD
101893-244 101893-246
3-[(4-Bromobenzyl)oxy]pyrrolidine hydrochloride
3-[(4-Bromobenzyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅BrClNO
MDL Number: MFCD13561217
CAS Number: 1220034-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR