4-{[(4-Bromobenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050836-500MG 050836-2.5G
101893-240EA 364.62 USD
101893-240 101893-242
4-{[(4-Bromobenzyl)oxy]methyl}piperidine hydrochloride
4-{[(4-Bromobenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13561216
CAS Number: 1220034-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR