3-{[(4-Bromobenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050835-500MG 050835-2.5G
101893-236EA 364.62 USD
101893-236 101893-238
3-{[(4-Bromobenzyl)oxy]methyl}piperidine hydrochloride
3-{[(4-Bromobenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13561215
CAS Number: 1220019-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR