3-[(3-Bromobenzyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050832-500MG 050832-2.5G
101893-224EA 364.62 USD
101893-224 101893-226
3-[(3-Bromobenzyl)oxy]pyrrolidine hydrochloride
3-[(3-Bromobenzyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅BrClNO
MDL Number: MFCD13561212
CAS Number: 1220034-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR