4-{[(3-Bromobenzyl)oxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050831-500MG 050831-2.5G
101893-220EA 364.62 USD
101893-220 101893-222
4-{[(3-Bromobenzyl)oxy]methyl}piperidine hydrochloride
4-{[(3-Bromobenzyl)oxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13561211
CAS Number: 1220034-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR