Methyl-4-[(4-piperidinylmethoxy)methyl]benzoate hydrochloride

Supplier: Matrix Scientific

050826-500MG 050826-2.5G
101893-200EA 364.62 USD
101893-200 101893-202
Methyl-4-[(4-piperidinylmethoxy)methyl]benzoate hydrochloride
Methyl-4-[(4-piperidinylmethoxy)methyl]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13561206
CAS Number: 1220020-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR