Methyl-4-[(3-piperidinylmethoxy)methyl]benzoate hydrochloride
Supplier: Matrix Scientific
050825-500MG
050825-2.5G
101893-196EA
364.62
USD
101893-196
101893-198
Methyl-4-[(3-piperidinylmethoxy)methyl]benzoate hydrochloride
Methyl-4-[(3-piperidinylmethoxy)methyl]benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₂ClNO₃ |
MDL Number:
MFCD13561205 CAS Number: 1220034-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...