Methyl-4-[(4-piperidinyloxy)methyl]benzoate hydrochloride
Supplier: Matrix Scientific
050824-500MG
050824-2.5G
101893-192EA
364.62
USD
101893-192
101893-194
Methyl-4-[(4-piperidinyloxy)methyl]benzoate hydrochloride
Methyl-4-[(4-piperidinyloxy)methyl]benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀ClNO₃ |
MDL Number:
MFCD13561204 CAS Number: 1220034-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...