Methyl-2-[(3-piperidinylmethoxy)methyl]benzoate hydrochloride

Supplier: Matrix Scientific

050820-500MG 050820-2.5G
101893-176EA 364.62 USD
101893-176 101893-178
Methyl-2-[(3-piperidinylmethoxy)methyl]benzoate hydrochloride
Methyl-2-[(3-piperidinylmethoxy)methyl]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13561200
CAS Number: 1220019-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR