Methyl-2-[(4-piperidinyloxy)methyl]benzoate hydrochloride

Supplier: Matrix Scientific

050819-500MG 050819-2.5G
101893-172EA 364.62 USD
101893-172 101893-174
Methyl-2-[(4-piperidinyloxy)methyl]benzoate hydrochloride
Methyl-2-[(4-piperidinyloxy)methyl]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD13561199
CAS Number: 1219976-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR