Methyl-2-[(3-piperidinyloxy)methyl]benzoate hydrochloride

Supplier: Matrix Scientific

050818-500MG 050818-2.5G
101893-168EA 364.62 USD
101893-168 101893-170
Methyl-2-[(3-piperidinyloxy)methyl]benzoate hydrochloride
Methyl-2-[(3-piperidinyloxy)methyl]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD13561198
CAS Number: 1220034-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR