2-Methoxy-4-propylphenyl-3-pyrrolidinyl ether hydrochloride

Supplier: Matrix Scientific

050817-500MG 050817-2.5G
101893-164EA 364.62 USD
101893-164 101893-166
2-Methoxy-4-propylphenyl-3-pyrrolidinyl ether hydrochloride
2-Methoxy-4-propylphenyl-3-pyrrolidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₂
MDL Number: MFCD13561197
CAS Number: 1220019-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR