2-Methoxy-4-propylphenyl-4-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050814-500MG 050814-2.5G
101893-152EA 364.62 USD
101893-152 101893-154
2-Methoxy-4-propylphenyl-4-piperidinyl ether hydrochloride
2-Methoxy-4-propylphenyl-4-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13561194
CAS Number: 1220020-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR