2-Methoxy-4-propylphenyl-3-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050813-500MG 050813-2.5G
101893-148EA 364.62 USD
101893-148 101893-150
2-Methoxy-4-propylphenyl-3-piperidinyl ether hydrochloride
2-Methoxy-4-propylphenyl-3-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13561193
CAS Number: 1220019-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR