Benzyl-4-(4-piperidinylmethoxy)benzoate hydrochloride

Supplier: Matrix Scientific

050811-500MG 050811-2.5G
101893-140EA 364.62 USD
101893-140 101893-142
Benzyl-4-(4-piperidinylmethoxy)benzoate hydrochloride
Benzyl-4-(4-piperidinylmethoxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₄ClNO₃
MDL Number: MFCD13561191
CAS Number: 1220034-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR