Benzyl-4-(4-piperidinyloxy)benzoate hydrochloride

Supplier: Matrix Scientific

050809-500MG 050809-2.5G
101893-132EA 364.62 USD
101893-132 101893-134
Benzyl-4-(4-piperidinyloxy)benzoate hydrochloride
Benzyl-4-(4-piperidinyloxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₂ClNO₃
MDL Number: MFCD13561189
CAS Number: 1220020-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR