Benzyl-4-(3-piperidinyloxy)benzoate hydrochloride

Supplier: Matrix Scientific

050808-500MG 050808-2.5G
101893-128EA 364.62 USD
101893-128 101893-130
Benzyl-4-(3-piperidinyloxy)benzoate hydrochloride
Benzyl-4-(3-piperidinyloxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₂ClNO₃
MW: 347.85 g/mol
MDL Number: MFCD13561188
CAS Number: 1219976-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR