3-[(2-Iodophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050805-500MG 050805-2.5G
101893-116EA 364.62 USD
101893-116 101893-118
3-[(2-Iodophenoxy)methyl]piperidine hydrochloride
3-[(2-Iodophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClINO
MDL Number: MFCD13561185
CAS Number: 1219976-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR