2-Iodophenyl-4-piperidinyl ether hydrochloride
Supplier: Matrix Scientific
050804-500MG
050804-2.5G
101893-112EA
364.62
USD
101893-112
101893-114
2-Iodophenyl-4-piperidinyl ether hydrochloride
2-Iodophenyl-4-piperidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClINO |
MDL Number:
MFCD13561184 CAS Number: 1220019-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...