2-Iodophenyl-3-piperidinyl ether hydrochloride
Supplier: Matrix Scientific
050803-500MG
050803-2.5G
101893-108EA
364.62
USD
101893-108
101893-110
2-Iodophenyl-3-piperidinyl ether hydrochloride
2-Iodophenyl-3-piperidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClINO |
MDL Number:
MFCD13561183 CAS Number: 1219976-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...