3-[(2-Nitrophenoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
050800-500MG
050800-2.5G
101893-096EA
364.62
USD
101893-096
101893-098
3-[(2-Nitrophenoxy)methyl]piperidine hydrochloride
3-[(2-Nitrophenoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂O₃ |
MDL Number:
MFCD13561181 CAS Number: 1219982-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...