4-(2-Nitrophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050799-500MG 050799-2.5G
101893-092EA 364.62 USD
101893-092 101893-094
4-(2-Nitrophenoxy)piperidine hydrochloride
4-(2-Nitrophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₃
MDL Number: MFCD09909466
CAS Number: 1072944-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR