2-Nitrophenyl-3-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050798-500MG 050798-2.5G
101893-088EA 364.62 USD
101893-088 101893-090
2-Nitrophenyl-3-piperidinyl ether hydrochloride
2-Nitrophenyl-3-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₃
MDL Number: MFCD09909465
CAS Number: 1220029-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR