3-(4-Isopropyl-3-methylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050797-500MG 050797-2.5G
101893-084EA 364.62 USD
101893-084 101893-086
3-(4-Isopropyl-3-methylphenoxy)pyrrolidine hydrochloride
3-(4-Isopropyl-3-methylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13561180
CAS Number: 1220019-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR