4-[(4-Isopropyl-3-methylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050796-500MG 050796-2.5G
101893-080EA 364.62 USD
101893-080 101893-082
4-[(4-Isopropyl-3-methylphenoxy)methyl]piperidine hydrochloride
4-[(4-Isopropyl-3-methylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13561179
CAS Number: 1219982-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR