4-(4-Isopropyl-3-methylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050794-500MG 050794-2.5G
101893-072EA 364.62 USD
101893-072 101893-074
4-(4-Isopropyl-3-methylphenoxy)piperidine hydrochloride
4-(4-Isopropyl-3-methylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13561177
CAS Number: 1219982-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR