3-(4-Isopropyl-3-methylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050793-500MG 050793-2.5G
101893-068EA 364.62 USD
101893-068 101893-070
3-(4-Isopropyl-3-methylphenoxy)piperidine hydrochloride
3-(4-Isopropyl-3-methylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13561176
CAS Number: 1220019-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR