3-(2-Bromo-4-fluorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050792-500MG 050792-2.5G
101893-064EA 364.62 USD
101893-064 101893-066
3-(2-Bromo-4-fluorophenoxy)pyrrolidine hydrochloride
3-(2-Bromo-4-fluorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂BrClFNO
MDL Number: MFCD13561175
CAS Number: 1220034-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR